missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl fluoride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Benzoyl fluoride is used as a mild fluorinating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzoyl fluoride is used as a mild fluorinating agent.
Solubility
Hydrolyses in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
Benzoyl fluoride is used as a mild fluorinating agent.
Solubility
Hydrolyses in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
Especificaciones
Melting Point | ∼-28°C |
Density | 1.14 |
Boiling Point | 159°C to 161°C |
Flash Point | 48°C (118°F) |
Odor | Pungent |
Refractive Index | 1.496 |
Quantity | 5 g |
UN Number | UN2920 |
Beilstein | 1904977 |
Sensitivity | Moisture sensitive |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'