missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Hydrazine, TRC

Product Code. 30331189
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331189 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331189 Supplier Toronto Research Chemicals Supplier No. TRCB20823525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoyldiazane (Benzohydrazide)
CAS 613-94-5
Purity Grade Notes HPLC
Molecular Formula C7 H8 N2 O
Quantity 25 g
Synonym Benzoyldiazane, Azelastine Hydrochloride Imp. A (EP), Benzohydrazide, Benzoic acid, hydrazide, Benzenecarboxylic acid hydrazide, Benzhydrazide, Benzoic hydrazide, Benzoylhydrazine, Hydrazine, benzoyl-, INHd 14, N-Benzoylhydrazine, NSC 644
InChI Formula InChI=1S/C7H8N2O/c8-9-7(10)6-4-2-1-3-5-6/h1-5H,8H2,(H,9,10)
SMILES NNC(=O)c1ccccc1
IUPAC Name benzohydrazide
Molecular Weight (g/mol) 136.15
Formula Weight 136.0637
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.