missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isocyanate, 90%, Tech.

Product Code. 10339170 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10339170

Brand: Thermo Scientific Acros 314280050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4461-33-0 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4461-33-0
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002032
InChI Key LURYMYITPCOQAU-UHFFFAOYSA-N
Synonym benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
PubChem CID 78210
IUPAC Name benzoyl isocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color White
Density 1.1700g/mL
Boiling Point 94°C to 96°C (21.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CONCO
Refractive Index 1.5504 to 1.5524
Quantity 5 g
Specific Gravity 1.17
Formula Weight 147.13
Percent Purity 90%
Grade Technical
Physical Form Solid
Chemical Name or Material Benzoyl isocyanate, tech., 90%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.