missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isocyanate, 90%, Tech.

Product Code. 10457333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10457333 1 g Glass bottle 1g
10339170 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457333 Supplier Thermo Scientific Acros Supplier No. 314280010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4461-33-0 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4461-33-0
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002032
InChI Key LURYMYITPCOQAU-UHFFFAOYSA-N
Synonym benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
PubChem CID 78210
IUPAC Name benzoyl isocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color White
Density 1.1700g/mL
Boiling Point 94°C to 96°C (21.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CONCO
Refractive Index 1.5504 to 1.5524
Quantity 1 g
Specific Gravity 1.17
Formula Weight 147.13
Percent Purity 90%
Grade Technical
Physical Form Solid
Chemical Name or Material Benzoyl isocyanate, tech., 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.