missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Peroxide - Contains 25% water, TRC

Product Code. 30345648
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345648 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345648 Supplier Toronto Research Chemicals Supplier No. TRCB218330250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoyl Peroxide
CAS 94-36-0
Purity Grade Notes HPLC
Molecular Formula C14 H10 O4
Quantity 250 mg
Synonym Benzoyl peroxide hydrous, Peroxide, dibenzoyl, Benzoyl peroxide (8CI), Dibenzoyl peroxide, 617P37, Abcat 40, Acetoxyl, Acne-Aid Cream, Acnegel, Acnezoyl, Akneroxide L, Aksil 5, Aztec BP 50FT, B 75W, BP, BP 50FT, BPO, BPO 50, Basiron, Benbel C, Benox 50, Benox A 80, Benoxyl, Benzac, Benzac W, Benzagel, Benzagel 10, Benzaknen, Benzashave, Benzoic peroxyanhydride, Benzoperoxide, Benzoyl superoxide, Brevoxyl, C 1A, C 1A (peroxide), CH 50, CH 50L, Cadet BPO, Cadet BPO 78W, Cadox 40E, Cadox B, Cadox B 40E, Cadox B 40ES, Cadox B 50P, Cadox B 75W, Cadox B-CH 50, Chaloxyd BP 50FT, Clear By Design, Debroxide, Degament H, Desanden, Desquam E, Desquam X, Diphenylglyoxal peroxide, Dry and Clear, Epi-Clear, Fivenox B 50G, G 20, Interox BP 50P1, Interox GZ-S, Link-Cup DBP, Loroxide, Lucidol, Lucidol (peroxide), Lucidol 40E, Lucidol 50P, Lucidol 70, Lucidol 75, Lucidol 75FP, Lucidol 75W, Lucidol 78, Lucidol 98, Lucidol B 50, Lucidol BT 50, Lucidol BW 50T, Lucidol CH 50, Lucidol CH 50L, Lucidol CH 50X, Lucidol G 20, Lucidol KL 50, Lucidol S 50, Lucidol W 40, Luperco AA, Luperco ANS, Luperco AST, Luperox 78, Luperox 98, Luperox A 40FP-EZ9, Luperox A 70S, Luperox A 75, Luperox A 75FP, Luperox A 98, Luperox ACP 35, Luperox AFR 40, Luperox ANS 50G, Luperox ATC 50, Luprox A 75C, NSC 671, NSC 675, Nericur, Niper BMT, Nyper B, Nyper BD, Nyper BMT, Nyper BMT 40, Nyper BMT 40SV, Nyper BMT-K, Nyper BO, Nyper BO-Y, Nyper BS, Nyper BW, Nyper F, Nyper FF, Nyper FF-K, Nyper NS, Oxy-5, Oxy-L, Oxycare 42, Oxylite, PEROX-B 75, PEROX-B 95, PHisoAc BP, Panoxyl, Perkadox 20S, Abcat 40, Acetoxyl, Acne-Aid Cream, Acnegel, Acnezoyl, Akneroxide L, Aksil 5, Aztec BP 50FT, B 75W, BP 50FT, BPO, BPO 50, Basiron, Benbel C, Benox 50, Benox A 80, Benoxyl, Benzac, Benzac W, Benzagel, Benzagel 10, Benzaknen, Benzashave, Benzoperoxide, Benzoyl Superoxide, Brevoxyl, C 1A, C 1A (peroxide), CH 50, CH 50L, Cadet BPO, Cadet BPO 78W, Cadox 40E, Cadox B, Cadox B 40E, Cadox B 40ES, Cadox B 50P, Cadox B 75W, Cadox B-CH 50, Chaloxyd BP 50FT, Clear By Design, Debroxide, Degament H, Desanden, Desquam E, Desquam X, Dibenzoyl peroxide, Diphenylglyoxal peroxide, Dry and Clear, Epi-Clear, Fivenox B 50G, G 20, Interox BP 50P1, Interox GZ-S, Link-Cup DBP, Loroxide, Lucidol, Lucidol (peroxide), Lucidol 40E, Lucidol 50P, Lucidol 70, Lucidol 75, Lucidol 75FP, Lucidol 75W, Lucidol 78, Lucidol 98Perkadox 33, Perkadox BT 50, Perkadox BTW 50, Perkadox CH 50, Perkadox CH 50L, Perkadox CH 50X, Perkadox L 40RPS, Perkadox L 50S, Perkadox L 50S-PS, Perkadox L-DFG, Perkadox L-W 40, Perkadox L-W 40TCP, Perkadox L-W 75, Perkadox PM 50S, Peroxan BP 50PF1, Peroxan BP 50SE, Peroxan BP Powder 50W, Peroxyderm, Persa-Gel, Persadox, Preoxydex, Sanoxit, Sanperox BPO, SikaFast 5200B, Solugel, Solugel (peroxide), Superox 46-750, Superox 744, TC 1, TC 1 (peroxide), Theraderm, Triaz, Vanoxide, Varox ANS, W 75, W 75 (peroxide), Xerac BP 10, Xerac BP 5, Abcat 40, Acetoxyl, Acne-Aid Cream, Acnegel, Acnezoyl, Akneroxide L, Aksil 5, Aztec BP 50FT, B 75W, BP 50FT, BPO, BPO 50, Basiron, Benbel C, Benox 50, Benox A 80, Benoxyl, Benzac, Benzac W, Benzagel, Benzagel 10, Benzaknen, Benzashave, Benzoperoxide, Benzoyl Superoxide, Brevoxyl, C 1A, C 1A (peroxide), CH 50, CH 50L, Cadet BPO, Cadet BPO 78W, Cadox 40E, Cadox B, Cadox B 40E, Cadox B 40ES, Cadox B 50P, Cadox B 75W, Cadox B-CH 50, Chaloxyd BP 50FT, Clear By Design, Debroxide, Degament H, Desanden, Desquam E, Desquam X, Dibenzoyl peroxide, Diphenylglyoxal peroxide, Dry and Clear, Epi-Clear, Fivenox B 50G, G 20, Interox BP 50P1, Interox GZ-S, Link-Cup DBP, Loroxide, Lucidol, Lucidol (peroxide), Lucidol 40E, Lucidol 50P, Lucidol 70, Lucidol 75, Lucidol 75FP, Lucidol 75W, Lucidol 78, Lucidol 98
InChI Formula InChI=1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H
SMILES O=C(OOC(=O)c1ccccc1)c2ccccc2
IUPAC Name benzoyl benzenecarboperoxoate
Molecular Weight (g/mol) 242.23
Formula Weight 242.0579
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.