missing translation for 'onlineSavingsMsg'
Learn More

Benzoylacetonitrile, 98+%

Product Code. 10627464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627464 25 g Glass bottle 25g
10599462 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 10627464 Lieferant Thermo Scientific Acros Lieferanten-Nr. 402000250
 Restricted Item
Estimated Shipment: 09-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 614-16-4 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 614-16-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001942
InChI Key ZJRCIQAMTAINCB-UHFFFAOYSA-N
Synonym benzoylacetonitrile, 2-cyanoacetophenone, phenacyl cyanide, 3-oxo-3-phenyl-propionitrile, cyanomethyl phenyl ketone, acetonitrile, benzoyl, cyanoacetophenone, 3-phenyl-3-ketopropionitrile, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile
PubChem CID 64799
ChEBI CHEBI:51855
IUPAC Name 3-oxo-3-phenylpropanenitrile
SMILES O=C(CC#N)C1=CC=CC=C1

Spezifikation

CAS Min % 98.0
CAS Max % 100.0
Melting Point 78.0°C to 82.0°C
Boiling Point 160.0°C (10.0 mmHg)
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5COCH2CN
Quantity 25 g
Beilstein 10, 680
Solubility Information Solubility in water: slightly soluble
Formula Weight 145.16
Percent Purity 98+%
Chemical Name or Material Benzoylacetonitrile, ≥98%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Benzoylacetonitrile
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.