missing translation for 'onlineSavingsMsg'
Learn More

Benzoylcholine iodide, 98+%, Thermo Scientific™

Product Code. 11450497
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11450497

Brand: Thermo Scientific Alfa Aesar B22387.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17518-43-3
Molecular Formula C12H18INO2
Molecular Weight (g/mol) 335.19
MDL Number MFCD00038726
InChI Key XILQCEGWGBSDSH-UHFFFAOYSA-M
Synonym benzoylcholine iodide, benzoylcholineiodide, 2-benzoyloxy-n,n,n-trimethylethanaminium iodide, 2-benzoyloxyethyl trimethylammonium iodide, ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide, ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide 1:1, 2-benzoyloxy ethyl trimethylazanium iodide, choline, iodide benzoate, choline, benzoate, acmc-1buu4
PubChem CID 87143
IUPAC Name 2-benzoyloxyethyl(trimethyl)azanium;iodide
SMILES [I-].C[N+](C)(C)CCOC(=O)C1=CC=CC=C1

Specifications

Melting Point ∼245°C (decomposition)
Linear Formula C6H5CO2CH2CH2N(CH3)3I
Quantity 1 g
Beilstein 3576708
Sensitivity Hygroscopic; Light sensitive
Formula Weight 335.19
Percent Purity ≥98%
Chemical Name or Material Benzoylcholine iodide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.