missing translation for 'onlineSavingsMsg'
Learn More

Benzoylcholine iodide, 98+%, Thermo Scientific™

Product Code. 11470497
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470497 25 g 25g
11450497 1 g 1g
11460497 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470497 Supplier Thermo Scientific Alfa Aesar Supplier No. B22387.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17518-43-3
Molecular Formula C12H18INO2
Molecular Weight (g/mol) 335.19
MDL Number MFCD00038726
InChI Key XILQCEGWGBSDSH-UHFFFAOYSA-M
Synonym benzoylcholine iodide, benzoylcholineiodide, 2-benzoyloxy-n,n,n-trimethylethanaminium iodide, 2-benzoyloxyethyl trimethylammonium iodide, ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide, ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide 1:1, 2-benzoyloxy ethyl trimethylazanium iodide, choline, iodide benzoate, choline, benzoate, acmc-1buu4
PubChem CID 87143
IUPAC Name 2-benzoyloxyethyl(trimethyl)azanium;iodide
SMILES [I-].C[N+](C)(C)CCOC(=O)C1=CC=CC=C1

Specifications

Melting Point ∼245°C (decomposition)
Linear Formula C6H5CO2CH2CH2N(CH3)3I
Quantity 25 g
Beilstein 3576708
Sensitivity Hygroscopic; Light sensitive
Formula Weight 335.19
Percent Purity ≥98%
Chemical Name or Material Benzoylcholine iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.