missing translation for 'onlineSavingsMsg'
Learn More

Benzthiazide, TRC

Product Code. 30137118
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137118 1 g 1g
30008860 50 mg 50mg
30012546 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137118 Supplier Toronto Research Chemicals Supplier No. TRCB2099251G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-33-8
Molecular Formula C15 H14 Cl N3 O4 S3
Molecular Weight (g/mol) 431.94
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 3-[(benzylthio)methyl]-6-chloro-, 1,1-dioxide (6CI,8CI), 3-Benzylthiomethyl-6-chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, 3-Benzylthiomethyl-6-chloro-7-sulfamoyl-1,2,4-benzothiadiazine 1,1-dioxide, 3-Benzylthiomethyl-6-chloro-7-sulfamyl-1,2,4-benzothiadiazine-1,1-dioxide, 3-Benzylthiomethyl-6-chloro-7-sulfamyl-2H-1,2,4-benzothiadiazine 1,1-dioxide, 3-[(Benzylthio)methyl]-6-chloro-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide, Aquatag, Benzothiazide, Benzthiazide, Dihydrex, Diucen, Diucene, Edemex, ExNa, Exosalt, Fovane, Freeuril, HyDrine, Lemazide, P 1393, Pfizer 1393, Proaqua, Regulon, Urese, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3-[[(phenylmethyl)thio]methyl]-, 1,1-dioxide
IUPAC Name 3-(benzylsulfanylmethyl)-6-chloro-1,1-dioxo-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(cc1Cl)N=C(CSCc3ccccc3)NS2(=O)=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H14ClN3O4S3/c16-11-6-12-14(7-13(11)25(17,20)21)26(22,23)19-15(18-12)9-24-8-10-4-2-1-3-5-10/h1-7H,8-9H2,(H,18,19)(H2,17,20,21)
Formula Weight 430.9835
Chemical Name or Material Benzthiazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.