missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2,2,2-Trichloroacetimidate, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | Benzyl 2,2,2-Trichloroacetimidate |
| CAS | 81927-55-1 |
| Molecular Formula | C9H8Cl3NO |
| Quantity | 100 g |
| Synonym | 2,2,2-Trichloroethanimidic Acid Phenylmethyl Ester, 2,2,2-Trichloroacetimidic Acid Benzyl Ester, Benzyl Trichloroacetimidate, O-Benzyl 2,2,2-Trichloroacetimidate |
| InChI Formula | InChI=1S/C9H8Cl3NO/c10-9(11,12)8(13)14-6-7-4-2-1-3-5-7/h1-5,13H,6H2 |
| SMILES | ClC(Cl)(Cl)C(=N)OCc1ccccc1 |
| IUPAC Name | benzyl 2,2,2-trichloroethanimidate |
| Molecular Weight (g/mol) | 252.52 |
| Formula Weight | 250.9671 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction