missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5-dimethyl-n-tetradecylammonium Bromide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Benzyl-2,3,4,5,6-d5-dimethyl-n-tetradecylammonium Bromide |
| CAS | 1515861-68-3 |
| Molecular Formula | C23 D5 H37 N . Br |
| Quantity | 0.05 g |
| Synonym | Benzene-2,3,4,5,6-d5-methanaminium, N,N-dimethyl-N-tetradecyl-, bromide (1:1) (ACI), N-Tetradecyl-N,N-dimethylbenzene-d5-methanaminium bromide, (Benzyl-2,3,4,5-d5)dimethyltetradecylammonium bromide, Benzyldimethyltetradecylammonium-D5 (phenyl-D5) Bromide |
| InChI Formula | InChI=1 S/C23H42N.BrH/c1-4-5-6-7-8-9-10-11-12-13-14-18-21-24(2,3)22-23-19-16-15-17-20-23;/h15-17,19-20 H,4-14,18,21-22H2,1-3H3;1 H/q+1;/p-1/i15D,16 D,17 D,19 D,20 D; |
| SMILES | [Br-].[2 H]c1c([2 H])c([2 H])c(C[N+](C)(C)CCCCCCCCCCCCCC)c([2 H])c1[2 H] |
| IUPAC Name | dimethyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]-tetradecylazanium;bromide |
| Molecular Weight (g/mol) | 417.5211 |
| Formula Weight | 416.2814 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction