missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-bromoacetate, 96%

Product Code. 15199253 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 15199253

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5437-45-6 | C9H9BrO2 | 229.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5437-45-6
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.08
MDL Number MFCD00000190
InChI Key JHVLLYQQQYIWKX-UHFFFAOYSA-N
Synonym benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
PubChem CID 62576
IUPAC Name benzyl 2-bromoacetate
SMILES C1=CC=C(C=C1)COC(=O)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4400g/mL
Boiling Point 166.0°C to 170.0°C (22.0 mmHg)
Flash Point >109°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5430 to 1.5450
Quantity 1 kg
Specific Gravity 1.44
Solubility Information Solubility in water: insoluble.
Formula Weight 229.08
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Benzyl 2-bromoacetate, 96%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.