missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate, ≥95%, Thermo Scientific™

Product Code. 10690656
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10690656

Brand: Thermo Scientific Maybridge CC13409DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 135829-04-8
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.327
MDL Number MFCD02682013
InChI Key QSMOQTIGILELKY-UHFFFAOYSA-N
Synonym benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline
PubChem CID 2776402
IUPAC Name benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate
SMILES C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 283.33
Percent Purity 95%
Chemical Name or Material Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.