missing translation for 'onlineSavingsMsg'
Learn More

Benzyl (2S,3R)-(+)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, 98%, Thermo Scientific™

Product Code. 10526674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10526674 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10526674 Supplier Thermo Scientific Acros Supplier No. 398670010
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 105228-46-4
Melting Point 205°C to 207°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C24H21NO4
MDL Number MFCD00074956
Quantity 1 g
Synonym 2s,3r-benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate, benzyl 2s,3r-+-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, benzyl 2s,3r-6-oxo-2,3-diphenylmorpholine-4-carboxylate, 2s,3r-n-cbz-6-oxo-2,3-diphenylmorpholine, pubchem18127, 2s,3r-+-n-z-6-oxo-2,3-diphenylmorpholine, 2s,3r-benzyl-6-oxo-2,3-diphenylmorpholine-4-carboxylate
InChI Key HECRUWTZAMPQOS-UHFFFAOYNA-N
SMILES O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name benzyl (2S,3R)-6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol) 387.44
PubChem CID 981233
Formula Weight 387.43
Percent Purity 98%
Chemical Name or Material Benzyl (2S, 3R)-(+)-6-oxo-2, 3-diphenyl-4-morpholinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl (2S,3R)-(+)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.