missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-[amino(thiocarbonyl)]piperidine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10481874
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10481874 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10481874 Supplier Thermo Scientific Maybridge Supplier No. MO08071CB
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 167757-46-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H18N2O2S
MDL Number MFCD09702422
Quantity 250 mg
Synonym benzyl 4-amino thiocarbonyl piperidine-1-carboxylate, piperidine-4-thiocarboxamide, n1-cbz protected, benzyl 4-aminocarbonothioyl piperidine-1-carboxylate, benzyl4-carbamothioylpiperidine-1-carboxylate, benzyl-4-carbamothioylpiperidine-1-carboxylate, 1-piperidinecarboxylicacid,4-aminothioxomethyl-,phenylmethyl ester
InChI Key NOXHDLSQVLNVJA-UHFFFAOYSA-N
SMILES C1CN(CCC1C(=S)N)C(=O)OCC2=CC=CC=C2
IUPAC Name benzyl 4-carbamothioylpiperidine-1-carboxylate
Molecular Weight (g/mol) 278.37
PubChem CID 18969329
Formula Weight 278.37
Percent Purity 97%
Chemical Name or Material Benzyl 4-[amino(thiocarbonyl)]piperidine-1-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.