missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate, 95%, Thermo Scientific™

Product Code. 10780174
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10780174 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10780174 Supplier Thermo Scientific Maybridge Supplier No. CC00094DA
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 167757-45-1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C14H18N2O3
MDL Number MFCD01568760
Quantity 1 g
Synonym benzyl 4-aminocarbonyl tetrahydro-1 2h-pyridinecarboxylate, 1-cbz-4-piperidinecarboxamide, 1-n-cbz-piperidine-4-carboxamide, 1-piperidinecarboxylic acid, 4-aminocarbonyl-, phenylmethyl ester, 4-carbamoyl-piperidine-1-carboxylic acid benzyl ester, n-z-isonipecotinamide, n-z-isonepocotinamide, buttpark 7550-02, ksc532o5b, 1-n-cbz-piperidine-4-carboxyamide
InChI Key UNJWOPBEKPMSGH-UHFFFAOYSA-N
SMILES C1CN(CCC1C(=O)N)C(=O)OCC2=CC=CC=C2
IUPAC Name benzyl 4-carbamoylpiperidine-1-carboxylate
Molecular Weight (g/mol) 262.309
PubChem CID 2776131
Formula Weight 262.31
Percent Purity 95%
Chemical Name or Material Benzyl 4-(aminocarbonyl)tetrahydro-1(2H)-pyridinecarboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.