missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-(bromomethyl)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Thermo Scientific™

Product Code. 10191252
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10191252 250 mg 250mg
10242972 1 g 1g
10392055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10191252 Supplier Thermo Scientific Maybridge Supplier No. CC07508CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 159275-17-9
Molecular Formula C14H18BrNO2
Molecular Weight (g/mol) 312.207
MDL Number MFCD02681989
InChI Key XJHKDSZGAWXUTB-UHFFFAOYSA-N
Synonym benzyl 4-bromomethyl piperidine-1-carboxylate, benzyl 4-bromomethyl tetrahydro-1 2h-pyridinecarboxylate, 1-cbz-4-bromomethyl piperidine, n-cbz-4-bromomethyl piperidine, phenylmethyl 4-bromomethyl piperidinecarboxylate, 1-piperidinecarboxylic acid, 4-bromomethyl-, phenylmethyl ester, benzyloxycarbonylbromomethylpiperidine, n-benzyloxycarbonyl-4-bromomethyl piperidine, benzyl 4-bromoethyl tetrahydro-1 2h-pyridinecarboxylate, benzyl 4-bromomethyl tetrahydro-1 2h pyridinecarboxylate
PubChem CID 2776274
IUPAC Name benzyl 4-(bromomethyl)piperidine-1-carboxylate
SMILES C1CN(CCC1CBr)C(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 312.21
Percent Purity 97%
Chemical Name or Material Benzyl 4-(bromomethyl)tetrahydro-1(2H)-pyridinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.