missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-oxo-1-piperidinecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 10376504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10376504 5 mL Glass bottle 5mL
10411114 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376504 Supplier Thermo Scientific Acros Supplier No. 425960050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 19099-93-5
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00673144
InChI Key VZOVOHRDLOYBJX-UHFFFAOYSA-N
Synonym 1-cbz-4-piperidone, n-benzyloxycarbonyl-4-piperidone, 1-cbz-piperidin-4-one, n-cbz-4-piperidone, benzyl 4-oxo-1-piperidinecarboxylate, 1-benzyloxycarbonyl-4-piperidinone, 1-z-4-piperidone, 1-piperidinecarboxylic acid, 4-oxo-, phenylmethyl ester, 1-carbobenzoxy-4-piperidone, 1-benzyloxycarbonyl-4-piperidone
PubChem CID 643496
IUPAC Name benzyl 4-oxopiperidine-1-carboxylate
SMILES C1CN(CCC1=O)C(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Density 1.1850g/mL
Boiling Point 114°C to 140°C (0.3 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 1.185
Formula Weight 233.27
Percent Purity 97%
Physical Form Liquid or Solid
Chemical Name or Material Benzyl 4-oxo-1-piperidinecarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl 4-oxo-1-piperidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.