missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 5-amino-1H-pyrazole-4-carboxylate, 98+%

Product Code. 11324805
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324805 1 g 1g
11334805 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324805 Supplier Thermo Scientific Alfa Aesar Supplier No. L17859.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 32016-28-7 | C11H11N3O2 | 217.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32016-28-7
Molecular Formula C11H11N3O2
Molecular Weight (g/mol) 217.23
MDL Number MFCD03426023
InChI Key QBEUXDWEKIOSIL-UHFFFAOYSA-N
Synonym benzyl 5-aminopyrazole-4-carboxylate, benzyl 3-amino-2h-pyrazole-4-carboxylate, acmc-20alpl, phenylmethyl 5-aminopyrazole-4-carboxylate, 3-amino-1h-pyrazole-4-carboxylic acid benzyl ester, 1h-pyrazole-4-carboxylicacid, 3-amino-, phenylmethyl ester
PubChem CID 4360500
IUPAC Name benzyl 5-amino-1H-pyrazole-4-carboxylate
SMILES NC1=C(C=NN1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 791498
Formula Weight 217.23
Percent Purity ≥98%
Chemical Name or Material Benzyl 5-amino-1H-pyrazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.