missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol

Product Code. 11366199
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366199 10 g 10g
11369268 50 g 50g
11359268 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366199 Supplier Thermo Scientific Alfa Aesar Supplier No. 043204.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Preparation of high refractive index polyacrylates | CAS: 2495-35-4 | C10H10O2 | 162.19 g/mol

Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2495-35-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00048147
InChI Key GCTPMLUUWLLESL-UHFFFAOYSA-N
Synonym benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
PubChem CID 75617
IUPAC Name benzyl prop-2-enoate
SMILES C=CC(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Density 1.06 g/mL
Boiling Point 111°C (8 mmHg)
Linear Formula C6H5CH2CO2CH=CH2
Refractive Index 1.518
Quantity 10 g
UN Number UN3082
Solubility Information Miscible with organic solvents and water.
Formula Weight 162.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzyl acrylate, Stabilized with ≈150ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.