missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Alcohol, TRC

Product Code. 30455509
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455509 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455509 Supplier Toronto Research Chemicals Supplier No. TRCT536635100G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl alcohol
CAS 100-51-6
Purity Grade Notes HPLC
Molecular Formula C7 H8 O
Quantity 100 g
Synonym Benzyl alcohol (8CI), (Hydroxymethyl)benzene, Benzenecarbinol, Benzylic alcohol, NSC 8044, Phenylcarbinol, Phenylmethanol, Phenylmethyl alcohol, Sunmorl BK 20, TB 13G, α-Hydroxytoluene, α-Toluenol
InChI Formula InChI=1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2
SMILES OCc1ccccc1
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
Formula Weight 108.0575
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.