missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Benzoate, TRC

Product Code. 30137286
Change view
Click to view available options
Quantity:
1 g
25 g
50 g
Unit Size:
1g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137286 50 g 50g
17902395 25 g 25g
17912935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137286 Supplier Toronto Research Chemicals Supplier No. TRCB23095050G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-51-4
Molecular Formula C14 H12 O2
Molecular Weight (g/mol) 212.24
Synonym Benzoic acid, benzyl ester (6CI,8CI), Ascabin, Ascabiol, Benylate, Benzyl benzenecarboxylate, Benzyl benzoate, Benzyl phenylformate, Benzylets, Benzyloxy phenyl ketone, Colebenz, NSC 8081, Nicca Sunsolt LM 7EX, Novoscabin, Pelemol B66, Peruscabin, Phenylmethyl benzoate, Scabagen, Scabanca, Scabcare BB, Scabide, Scabiozon, Scobenol, Vanzoate, Venzonate
IUPAC Name benzyl benzoate
SMILES O=C(OCc1ccccc1)c2ccccc2

Specifications

InChI Formula InChI=1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2
Formula Weight 212.0837
Percent Purity >95%
Quantity 50 g
Chemical Name or Material Benzyl Benzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.