missing translation for 'onlineSavingsMsg'
Learn More

Benzyl carbamate, 98%, BLD Pharm™

Product Code. 31094037
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31094037 10 g
31105743 100 g
31100567 25 g
31103612 1 kg
31107954 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31094037 Supplier BLD Pharm Supplier No. BD11612410g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 621-84-1
Molecular Formula C8H9NO2
MDL Number MFCD00007965
Quantity 10 g
SMILES NC(OCC1=CC=CC=C1)=O
Molecular Weight (g/mol) 151.16
Percent Purity 98%
Chemical Name or Material Benzyl carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.