missing translation for 'onlineSavingsMsg'
Learn More

Benzyl carbazate, 98+%

Product Code. 11489903
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11489903

Brand: Thermo Scientific Alfa Aesar A13005.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5331-43-1 | C8H10N2O2 | 166.18 g/mol

Benzyl carbazate is used as a reagent for preparation of benzyloxycarbonyl (Z, Cbz) hydrazides of carboxylic acids, particularly N-protected amino acids. Subsequent hydrogenolysis gives the hydrazide which can be converted to the azide for coupling with a further amino acid residue by the azide method. It is also used in the preparation of simple acyl or sulfonyl hydrazides avoiding diacylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl carbazate is used as a reagent for preparation of benzyloxycarbonyl (Z, Cbz) hydrazides of carboxylic acids, particularly N-protected amino acids. Subsequent hydrogenolysis gives the hydrazide which can be converted to the azide for coupling with a further amino acid residue by the azide method. It is also used in the preparation of simple acyl or sulfonyl hydrazides avoiding diacylation.

Solubility
Soluble in water (slightly), methanol and DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle under inert gas. Protect from moisture. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5331-43-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00041890
InChI Key RXUBZLMIGSAPEJ-UHFFFAOYSA-N
Synonym benzyl carbazate, benzyl hydrazinecarboxylate, hydrazinecarboxylic acid, phenylmethyl ester, carbobenzoxyhydrazide, carbazic acid, benzyl ester, benzyloxycarbonyl hydrazide, benzylcarbazate, benzyloxy carbonyl hydrazine, n-cbz-hydrazine, unii-h7gm2h55tc
PubChem CID 79242
IUPAC Name benzyl N-aminocarbamate
SMILES C1=CC=C(C=C1)COC(=O)NN

Specifications

Melting Point 66°C to 70°C
Quantity 25 g
Beilstein 1952982
Solubility Information Soluble in water (slightly),methanol and DMSO.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Benzyl carbazate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.