missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Carbazate, TRC

Product Code. 30330624
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330624 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330624 Supplier Toronto Research Chemicals Supplier No. TRCB23362010G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Carbazate
CAS 5331-43-1
Molecular Formula C8H10N2O2
Quantity 10 g
Synonym Carbazic Acid Benzyl Ester, Benzyl Carbazate, Benzyl Hydrazinecarboxylate, Benzyl Hydrazinocarboxylate, Benzyloxycarbonyl Hydrazide, Carbobenzoxyhydrazide, Carbobenzyloxyhydrazine, Hydrazincarboxylic Acid Benzyl Ester, Hydrazinecarboxylic Acid Benzyl Ester, NSC 2287, [(Benzyloxy)carbonyl]hydrazine
InChI Formula InChI=1S/C8H10N2O2/c9-10-8(11)12-6-7-4-2-1-3-5-7/h1-5H,6,9H2,(H,10,11)
SMILES NNC(=O)OCc1ccccc1
IUPAC Name benzyl N-aminocarbamate
Molecular Weight (g/mol) 166.18
Formula Weight 166.0742
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.