missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloride, TRC

Product Code. 30347829
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30347829 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30347829

Brand: Toronto Research Chemicals TRCB2336755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material alpha-Chlorotoluene (Benzylchloride)
CAS 100-44-7
Purity Grade Notes HPLC
Molecular Formula C7 H7 Cl
Quantity 5 g
Synonym Toluene, α-chloro- (8CI), (Chloromethyl)benzene, 1-Chloromethylbenzene, Benzyl chloride, Bnol, Chlorophenylmethane, NSC 8043, Phenylmethyl chloride, Tolyl chloride, α-Chlorotoluene, ω-Chlorotoluene
InChI Formula InChI=1S/C7H7Cl/c8-6-7-4-2-1-3-5-7/h1-5H,6H2
SMILES ClCc1ccccc1
IUPAC Name chloromethylbenzene
Molecular Weight (g/mol) 126.58
Formula Weight 126.0236
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.