missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™

Product Code. 10119864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10119864 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10119864 Supplier Thermo Scientific Acros Supplier No. 428901000
  • ONLINE EXCLUSIVE PRICE
    34.30 €
    33.30 € / 100mL
    Save 1.00 €
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl chloroformate
CAS 501-53-1, 108-88-3, 100-44-7
CAS Min % 49.83, 44.26
CAS Max % 55.74, 50.17
Color Yellow to Pink
Density 1.0100g/mL
Flash Point 7°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H7ClO2
Linear Formula C6H5CH2OOCCl
MDL Number MFCD00000640
Quantity 100 mL
Merck Index 15, 1801
Specific Gravity 1.01
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
Solubility Information Solubility in water: decomposes
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC(=O)Cl
IUPAC Name benzyl carbonochloridate
Molecular Weight (g/mol) 170.6
PubChem CID 10387
Formula Weight 170.6
Percent Purity 45 to 53 wt%
Physical Form Solution
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Benzyl chloroformate, 50 wt% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Flammable liquid Category 2
Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H304-May be fatal if swallowed and enters airways.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Toluene, Benzyl chloroformate, Benzyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.