missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 97 wt%, stabilized

Product Code. 10222870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10222870 2.5 kg Glass bottle 2.5kg
10183580 5 g Glass bottle 5g
10771394 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10222870 Supplier Thermo Scientific Acros Supplier No. 152940025
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Benzyl chloroformate, 97 wt%, stabilized, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437 | CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-53-1 , 100-44-7
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Melting Point -20.0°C
Density 1.2000g/mL
Boiling Point 178.0°C to 180.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2CH2C6H5
Refractive Index 1.5160 to 1.5180
Quantity 2.5 kg
Beilstein 06,437
Fieser 01,109; 02,59; 15,22
Merck Index 15,1801
Specific Gravity 1.2
Solubility Information Solubility in water: reacts
Stabilizer 0.1% anhydrous sodium carbonate
Viscosity 2.36 mPa.s (20°C)
Formula Weight 170.6
Percent Purity 97 wt%
Physical Form Liquid
Chemical Name or Material Benzyl chloroformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.