missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cinnamate, 99%

Product Code. 11474117
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474117 500 g 500g
11464117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474117 Supplier Thermo Scientific Alfa Aesar Supplier No. A19550.36
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-41-3 | C16H14O2 | 238.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-41-3
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.29
MDL Number MFCD00004789,MFCD00004789,MFCD00004789
InChI Key NGHOLYJTSCBCGC-VAWYXSNFSA-N
Synonym benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z
PubChem CID 15558051
IUPAC Name benzyl (Z)-3-phenylprop-2-enoate
SMILES O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1

Specifications

Melting Point 34°C to 36°C
Density 1.11
Boiling Point 195°C to 200°C (5 mmHg)
Flash Point 180°C (356°F)
Quantity 500 g
Beilstein 2051339
Merck Index 14,1130
Formula Weight 238.29
Percent Purity 99%
Chemical Name or Material Benzyl cinnamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.