missing translation for 'onlineSavingsMsg'
Learn More

Benzyl dimethyl phosphonoacetate, 98%

Product Code. 10669414 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10669414 5 g Glass bottle 5g
10260033 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10669414 Supplier Thermo Scientific Acros Supplier No. 387370050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 57443-18-2 | C11H15O5P | 258.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57443-18-2
Molecular Formula C11H15O5P
Molecular Weight (g/mol) 258.21
MDL Number MFCD01872507
InChI Key QYLGNJMIOVHLQQ-UHFFFAOYSA-N
Synonym benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
PubChem CID 2773759
IUPAC Name benzyl 2-dimethoxyphosphorylacetate
SMILES COP(=O)(CC(=O)OCC1=CC=CC=C1)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Brown
Boiling Point 148.0°C to 150.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)2P(O)CH2C(O)OCH2C5H5
Refractive Index 1.5050 to 1.5070
Quantity 5 g
Formula Weight 258.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzyl dimethyl phosphonoacetate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl dimethyl phosphonoacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.