missing translation for 'onlineSavingsMsg'
Learn More

Benzyl ethyl malonate, tech. 85%

Product Code. 11456186
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456186 25 g 25g
11446186 5 g 5g
11466186 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456186 Supplier Thermo Scientific Alfa Aesar Supplier No. B23011.14
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 42998-51-6 | C12H14O4 | 222.24 g/mol

Benzyl ethyl malonate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl ethyl malonate is used as a pharmaceutical intermediate.

Solubility
Soluble in water 525.7 mg/L at 25°C. Soluble in alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42998-51-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009194
InChI Key CGNOCUSLPSCMLL-UHFFFAOYSA-N
Synonym benzyl ethyl malonate, benzylethyl malonate, malonic acid benzyl ethyl ester, ethyl benzylmalonate, acmc-20alpf, 1-benzyl 3-ethyl malonate #, ethyl phenylmethyl propanedioate, 1-benzyl 3-ethyl propanedioate, malonic acid 1-benzyl 3-ethyl ester, ethyl phenylmethyl propane-1,3-dioate
PubChem CID 562228
SMILES CCOC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Boiling Point 145°C (4 mmHg)
Refractive Index 1.497
Quantity 25 g
Solubility Information Soluble in water 525.7mg/L @ 25°C. Soluble in alcohol.
IUPAC Name 1-benzyl 3-ethyl propanedioate
Formula Weight 222.24
Percent Purity 85%
Grade Technical
Chemical Name or Material Benzyl ethyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.