missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Isocyanate, TRC

Product Code. 30333524
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333524 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333524 Supplier Toronto Research Chemicals Supplier No. TRCB27979525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Isocyanate
CAS 3173-56-6
Molecular Formula C8H7NO
Quantity 25 g
Synonym (Isocyanatomethyl)benzene, 1-(Isocyanatomethyl)benzene, Benzyl isocyanate, NSC 118415, Phenylmethyl isocyanate, alpha-Isocyanatotoluene
InChI Formula InChI=1S/C8H7NO/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2
SMILES O=C=NCc1ccccc1
IUPAC Name isocyanatomethylbenzene
Molecular Weight (g/mol) 133.15
Formula Weight 133.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.