missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isocyanide, 98%

Product Code. 10763583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10763583 1 g Glass bottle 1g
10622793 5 g Glass bottle 5g
11962411 25 g Glass bottle 25g
3 options
This item is not returnable. View return policy

Product Code. 10763583

Brand: Thermo Scientific Acros 158390010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10340-91-7 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10340-91-7
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00000004
InChI Key RIWNFZUWWRVGEU-UHFFFAOYSA-N
Synonym benzyl isocyanide, isocyanomethyl benzene, benzylisocyanide, benzene, isocyanomethyl, benzyl isonitrile, benzylimino methylidene, benzylisonitrile, benzylisonit-rile, isocyanomethylbenzol, isocyanomethyl-benzene
PubChem CID 82558
IUPAC Name isocyanomethylbenzene
SMILES [C-]#[N+]CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9620g/mL
Boiling Point 105.0°C to 106.0°C (75.0 mmHg)
Flash Point 79°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2NC
Refractive Index 1.5200 to 1.5220
Quantity 1 g
Beilstein 12,1041
Specific Gravity 0.962
Formula Weight 117.18
Percent Purity 98%
Chemical Name or Material Benzyl isocyanide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl isocyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.