missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isothiocyanate, 98%

Product Code. 10752084 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10752084 5 g Glass bottle 5g
10677181 25 g Glass bottle 25g
10378623 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10752084 Supplier Thermo Scientific Acros Supplier No. 402130050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-78-6 | C8H7NS | 149.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-78-6
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.22
MDL Number MFCD00004819
InChI Key MDKCFLQDBWCQCV-UHFFFAOYSA-N
Synonym benzyl isothiocyanate, isothiocyanatomethyl benzene, benzyl mustard oil, benzylisothiocyanate, benzylsenfoel, tromacaps, tromalyt, urogran, isothiocyanic acid, benzyl ester, benzyl-isothiocyanate
PubChem CID 2346
ChEBI CHEBI:17484
IUPAC Name isothiocyanatomethylbenzene
SMILES C1=CC=C(C=C1)CN=C=S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1200g/mL
Boiling Point 243.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2NCSC6H5
Refractive Index 1.6010 to 1.6040
Quantity 5 g
Specific Gravity 1.12
Formula Weight 149.22
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzyl isothiocyanate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.