missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, 99%

Product Code. 11384579
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384579 25 g 25g
11490571 100 g 100g
11400581 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384579 Supplier Thermo Scientific Alfa Aesar Supplier No. B20081.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100-53-8 | C7H8S | 124.20 g/mol

A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 1.053
Boiling Point 194°C to 195°C
Flash Point 70°C (158°F)
Assay Percent Range 0.99
Odor Stench
Refractive Index 1.576
Quantity 25 g
UN Number UN2810
Beilstein 605864
Sensitivity Air Sensitive
Merck Index 14,9322
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzyl mercaptan
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzenemethanethiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.