missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Mercaptan, TRC

Product Code. 30329666
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329666 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329666 Supplier Toronto Research Chemicals Supplier No. TRCB2855051G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Mercaptan
CAS 100-53-8
Molecular Formula C7 H8 S
Quantity 1 g
Synonym α-Toluenethiol (7CI,8CI), (Mercaptomethyl)benzene, Benzyl hydrosulfide, Benzyl mercaptan, Benzylthiol, Ethoxylated bisphenol A dimethacrylate-Benzyl mercaptan copolymer, GMP 800, NSC 41897, Phenylmethanethiol, Phenylmethyl mercaptan, Thiobenzyl alcohol, α-Mercaptotoluene, α-Toluolthiol, α-Tolyl mercaptan, Benzenemethanethiol
InChI Formula InChI=1S/C7H8S/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2
SMILES SCc1ccccc1
IUPAC Name phenylmethanethiol
Molecular Weight (g/mol) 124.2
Formula Weight 124.0347
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.