missing translation for 'onlineSavingsMsg'
Learn More

Benzyl N-(2-aminoethyl)carbamate hydrochloride, 97%, BLD Pharm™

Product Code. 30802272
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
250mg
25g
500g
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30802272 25 g 25g
30800057 250 mg 250mg
30796238 100 g 100g
30788127 1 g 1g
30800745 500 g 500g
30787284 10 g 10g
30791776 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 30802272 Supplier BLD Pharm Supplier No. BD11618325g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18807-71-1
Molecular Formula C10H15ClN2O2
MDL Number MFCD00270150
Quantity 25 g
SMILES O=C(OCC1=CC=CC=C1)NCCN.[H]Cl
Molecular Weight (g/mol) 230.69
Percent Purity 97%
Chemical Name or Material Benzyl N-(2-aminoethyl)carbamate hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.