missing translation for 'onlineSavingsMsg'
Learn More

Benzyl sulfoxide, 98+%, Thermo Scientific Chemicals

Product Code. 10531184 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10531184 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10531184 Supplier Thermo Scientific Acros Supplier No. 106175000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-08-9
Molecular Formula C14H14OS
Molecular Weight (g/mol) 230.33
MDL Number MFCD00004782
InChI Key HTMQZWFSTJVJEQ-UHFFFAOYSA-N
Synonym dibenzyl sulfoxide, benzyl sulfoxide, dibenzylsulfoxide, tardiol d, sulfoxide, dibenzyl, dibenzyl sulphoxide, sulfinylbis methylene dibenzene, bis phenylmethyl sulfoxide, nsc 55, benzene, 1,1'-sulfinylbis methylene bis
PubChem CID 12116
IUPAC Name benzylsulfinylmethylbenzene
SMILES O=S(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 130°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5CH2)2SO
Quantity 500 g
Beilstein 06, 456
Solubility Information Solubility in water: 0,3g/L (20°C). Other solubilities: soluble in ethanol and methanol
Formula Weight 230.32
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Benzyl sulfoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.