missing translation for 'onlineSavingsMsg'
Learn More

Benzyl thioacetimidate hydrochloride, 96%

Product Code. 15458317
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15458317 1 g 1g
15468317 5 g 5g
15478317 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15458317 Supplier Thermo Scientific Alfa Aesar Supplier No. H51844.03
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 32894-07-8 | C9H12ClNS | 201.712 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32894-07-8
Molecular Formula C9H12ClNS
Molecular Weight (g/mol) 201.712
MDL Number MFCD05865210
InChI Key OVQWHRFBVPDPCD-UHFFFAOYSA-N
Synonym s-benzylthioacetimidate, hydrochloride, benzyl thioacetimidate hydrochloride, benzyl thioacetimidate hcl, benzyl ethanimidothioate hydrochloride, 1-benzylsulfanyl ethanimine hydrochloride, acmc-20ajr8, s-benzylthioacetimidate hydrochloride, thioacetimidic acid benzyl ester; hydrochloride, benzyl ethanimidothioate-hydrogen chloride 1/1
PubChem CID 12626231
IUPAC Name benzyl ethanimidothioate;hydrochloride
SMILES CC(=N)SCC1=CC=CC=C1.Cl

Specifications

Quantity 1 g
Formula Weight 201.71
Percent Purity 96%
Chemical Name or Material Benzyl thioacetimidate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.