missing translation for 'onlineSavingsMsg'
Learn More

Benzylboronic acid pinacol ester, 96%

Product Code. 15475247
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475247 1 g 1g
15485247 5 g 5g
15495247 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475247 Supplier Thermo Scientific Alfa Aesar Supplier No. H32523.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 87100-28-5 | C13H19BO2 | 218.103 g/mol

Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylboronic acid pinacol ester finds application in Pd (0)-mediated [11 C] carbonylation reaction.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87100-28-5
Molecular Formula C13H19BO2
Molecular Weight (g/mol) 218.103
MDL Number MFCD05663841
InChI Key YCNQPAVKQPLZRS-UHFFFAOYSA-N
Synonym benzylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl, acmc-209qek, benzylboronic pinacol ester, benzyl boronic acid pinacol ester, benzylboronic acid, pinacol ester, 2-benzyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4,4,5,5-tetramethyl-2-phenylmethyl-1,3,2-dioxaborolane
PubChem CID 3864964
IUPAC Name 2-benzyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CC2=CC=CC=C2

Specifications

Density 0.98
Boiling Point 65°C (0.15 mmHg)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 218.1
Percent Purity 96%
Chemical Name or Material Benzylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.