missing translation for 'onlineSavingsMsg'
Learn More

Benzyldimethyl-n-dodecylammonium chloride, 98%

Product Code. 15476668
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476668 5 g 5g
15486668 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476668 Supplier Thermo Scientific Alfa Aesar Supplier No. H60196.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 139-07-1 | C21H38ClN | 339.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-07-1
Molecular Formula C21H38ClN
Molecular Weight (g/mol) 339.99
MDL Number MFCD00137276
InChI Key JBIROUFYLSSYDX-UHFFFAOYSA-M
Synonym benzyldimethyldodecylammonium chloride, benzododecinium chloride, n-benzyl-n,n-dimethyldodecan-1-aminium chloride, cequartyl a, catigene om, benzyldodecyldimethylammonium chloride, zephirol, dehyquart ldb, catiogen pan, vantoc cl
PubChem CID 8753
IUPAC Name benzyl-dodecyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1

Specifications

Melting Point 60°C
Quantity 5 g
UN Number UN3261
Beilstein 3921689
Formula Weight 339.99
Percent Purity 98%
Chemical Name or Material Benzyldimethyl-n-dodecylammonium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.