missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrochlorothiazide, TRC

Product Code. 30011614
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011614 100 mg 100mg
30005850 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011614 Supplier Toronto Research Chemicals Supplier No. TRCB279020100MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-50-6
Molecular Formula C14 H14 Cl N3 O4 S2
Molecular Weight (g/mol) 387.86
Synonym 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 6-chloro-3,4-dihydro-3-(phenylmethyl)-, 1,1-dioxide, 2H-1,2,4-Benzothiadiazine-7-sulfonamide, 3-benzyl-6-chloro-3,4-dihydro-, 1,1-dioxide (6CI,7CI,8CI), (±)-Benzylhydrochlorothiazide, Behyd, Benclortriazide, Benzclortriazide, Benzylhydrochlorothiazide, NSC 263220, Su 6227
IUPAC Name 3-benzyl-6-chloro-1,1-dioxo-3,4-dihydro-2H-1λ^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)c1cc2c(NC(Cc3ccccc3)NS2(=O)=O)cc1Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H14ClN3O4S2/c15-10-7-11-13(8-12(10)23(16,19)20)24(21,22)18-14(17-11)6-9-4-2-1-3-5-9/h1-5,7-8,14,17-18H,6H2,(H2,16,19,20)
Formula Weight 387.0114
Percent Purity >95%
Chemical Name or Material Benzylhydrochlorothiazide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.