missing translation for 'onlineSavingsMsg'
Learn More

Benzylmagnesium chloride, 1-2M in THF, Thermo Scientific Chemicals

Product Code. 15432967
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432967 50 mL 50mL
15442967 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432967 Supplier Thermo Scientific Alfa Aesar Supplier No. 087299.AD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6921-34-2
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.89
MDL Number MFCD00000469
InChI Key NRAFPLGJPPJUNB-UHFFFAOYSA-M
Synonym benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
PubChem CID 2733352
IUPAC Name magnesium;methanidylbenzene;chloride
SMILES Cl[Mg]CC1=CC=CC=C1

Specifications

Color Brown to Purple
Quantity 50 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Formula Weight 150.89
Concentration or Composition (by Analyte or Components) 1 to 2M in THF
Physical Form Liquid
Chemical Name or Material Benzylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.