missing translation for 'onlineSavingsMsg'
Learn More

Benzylmagnesium chloride, 1M in MeTHF

Product Code. 15488027
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488027 500 mL 500mL
15478027 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488027 Supplier Thermo Scientific Alfa Aesar Supplier No. H51154.AP
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 6921-34-2 | C7H7ClMg | 150.89 g/mol

Benzyl magnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl magnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is also used as organic chemical synthesis intermediate.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6921-34-2
Molecular Formula C7H7ClMg
Molecular Weight (g/mol) 150.89
MDL Number MFCD00000469
InChI Key NRAFPLGJPPJUNB-UHFFFAOYSA-M
Synonym benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
PubChem CID 2733352
IUPAC Name magnesium;methanidylbenzene;chloride
SMILES Cl[Mg]CC1=CC=CC=C1

Specifications

Quantity 500 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
Formula Weight 150.89
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Benzylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.