missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%

Product Code. 11450697
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450697 5 g 5g
11440697 1 g 1g
11460697 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450697 Supplier Thermo Scientific Alfa Aesar Supplier No. B22506.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 88568-95-0 | C13H18NO7P | 331.261 g/mol

(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88568-95-0
Molecular Formula C13H18NO7P
Molecular Weight (g/mol) 331.261
MDL Number MFCD00043304
InChI Key GSYSFVSGPABNNL-UHFFFAOYSA-N
Synonym methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate, +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, n-cbz-2-phosphonoglycine trimethyl ester, n-benzyloxy carbonyl, +/--cbz-alpha-phosphonoglycine trimethyl ester, methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate, +/--n-cbz-alpha-phosphonoglycine trimethyl ester, z-alpha-phosphonoglycine trimethyl ester, cbz-alpha-phosphono-dl-glycine trimethyl ester
PubChem CID 2734718
IUPAC Name methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
SMILES COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC

Specifications

Melting Point 76°C to 80°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 331.27
Percent Purity 97%
Chemical Name or Material (+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.