missing translation for 'onlineSavingsMsg'
Learn More

Benzylphosphonic acid, 97%

Product Code. 11461508
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461508 25 g 25g
11441508 1 g 1g
11451508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461508 Supplier Thermo Scientific Alfa Aesar Supplier No. A13850.14
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6881-57-8 | C7H7O3P | 170.11 g/mol

Structural mimic of phosphotyrosine

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6881-57-8
Molecular Formula C7H7O3P
Molecular Weight (g/mol) 170.11
MDL Number MFCD00039519
InChI Key OGBVRMYSNSKIEF-UHFFFAOYSA-L
Synonym benzyl-phosphonic acid, phosphonic acid, phenylmethyl, phenylmethylphosphonic acid, phosphonic acid, p-phenylmethyl, phenyl-methanephosphonic acid anion, acmc-1b2tf, phenylmethanephosphonic acid, alpha-toluenephosphonic acid, benzylphosphonic acid, bmk1-h2
PubChem CID 81312
IUPAC Name benzylphosphonic acid
SMILES [O-]P([O-])(=O)CC1=CC=CC=C1

Specifications

Melting Point 166°C to 172°C
Quantity 25 g
UN Number UN3261
Beilstein 2519318
Formula Weight 172.11
Percent Purity 97%
Chemical Name or Material Benzylphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.