missing translation for 'onlineSavingsMsg'
Learn More

(Benzylthio)acetone, 98%, Thermo Scientific Chemicals

Product Code. 11346666
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11346666 25 g 25g
11336666 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346666 Supplier Thermo Scientific Alfa Aesar Supplier No. L03814.14

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10230-69-0
Molecular Formula C10H12OS
Molecular Weight (g/mol) 180.265
MDL Number MFCD00026241
InChI Key OIEDQMIEPJIRFT-UHFFFAOYSA-N
Synonym 1-benzylthio acetone, 1-benzylthio-2-propanone, benzylthio acetone, .alpha.-benzylthio acetone, 1-benzylsulfanyl propan-2-one, 1-phenylmethylthio acetone, 1-benzylsulfanyl acetone, benzylthio-2-propanone, acmc-20ap9q, alpha-benzylthio acetone
PubChem CID 82472
IUPAC Name 1-benzylsulfanylpropan-2-one
SMILES CC(=O)CSCC1=CC=CC=C1

Specifications

Density 1.09
Boiling Point 155°C to 156°C (17 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.558
Quantity 25 g
Beilstein 1866268
Formula Weight 180.27
Percent Purity 98%
Chemical Name or Material (Benzylthio)acetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.