missing translation for 'onlineSavingsMsg'
Learn More

Benzyltributylammonium chloride, 98+%

Product Code. 10072300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10072300 500 g Glass bottle 500g
10164440 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10072300 Supplier Thermo Scientific Acros Supplier No. 186155000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23616-79-7 | C19H34ClN | 311.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23616-79-7
Molecular Formula C19H34ClN
Molecular Weight (g/mol) 311.94
MDL Number MFCD00011849
InChI Key VJGNLOIQCWLBJR-UHFFFAOYSA-M
Synonym benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
PubChem CID 159952
IUPAC Name benzyl(tributyl)azanium;chloride
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 155°C to 163°C
Color White to Ivory
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5CH2N[(CH2)3CH3]3Cl
Quantity 500 g
Fieser 06,41
Solubility Information Solubility in water: freely soluble in water
Formula Weight 311.94
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material Benzyltributylammonium chloride, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • Benzyltributylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.