missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyltributylammonium Chloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Benzyltributylammonium Chloride |
| CAS | 23616-79-7 |
| Molecular Formula | C19H34ClN |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C19H34N.ClH/c1-4-7-15-20(16-8-5-2,17-9-6-3)18-19-13-11-10-12-14-19;/h10-14H,4-9,15-18H2,1-3H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CCCC[N+](CCCC)(CCCC)Cc1ccccc1 |
| IUPAC Name | benzyl(tributyl)azanium;chloride |
| Molecular Weight (g/mol) | 311.93 |
| Formula Weight | 311.238 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction