missing translation for 'onlineSavingsMsg'
Learn More

Benzyltrimethylammonium Hydroxide (~35% wt % in H2O), TRC

Product Code. 30330454
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30330454 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330454 Supplier Toronto Research Chemicals Supplier No. TRCB31616850G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Benzyltrimethylammonium Hydroxide
CAS 100-85-6
Molecular Formula C10H17NO
Quantity 50 g
Synonym Benzyltrimethylammonium Hydroxide, BTMAH 40, N,N,N-Trimethyl-N-benzylammonium Hydroxide, N,N,N-Trimethylbenzenemethanaminium Hydroxide, N-Benzyltrimethylammonium Hydroxide, Sumquat 2311, Trimethylbenzylammonium Hydroxide, Triton B
InChI Formula InChI=1S/C10H16N.H2O/c1-11(2,3)9-10-7-5-4-6-8-10;/h4-8H,9H2,1-3H3;1H2/q+1;/p-1
SMILES [OH-].C[N+](C)(C)Cc1ccccc1
IUPAC Name benzyl(trimethyl)azanium;hydroxide
Molecular Weight (g/mol) 167.25
Formula Weight 167.131
Concentration or Composition (by Analyte or Components) 35 %
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.